
Zidovudine Related Compound D

Synonym(s):
Thymidine, 1-(2-Deoxy-β-D-ribofuranosyl)-5-methyluracil, 1-(2-Deoxy-β-D-ribofuranosyl)thymine, 2′-Deoxythymidine, Thymine deoxyriboside, dT
Empirical Formula (Hill Notation):
C10H14N2O5
CAS Number:
Molecular Weight:
242.23
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2