
Valganciclovir hydrochloride hydrate

Synonym(s):
2-[(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)methoxy]-3-hydroxypropyl (2S)-2-amino-3-methylbutanoate hydrochloride hydrate, L-Valine, 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]-3-hydroxypropyl ester, monohydrochloride hydrate
Empirical Formula (Hill Notation):
C14H22N6O5·HCl · xH2O
CAS Number:
Molecular Weight:
390.82 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing