
Triirondodecacarbonyl

Synonym(s):
Dodecacarbonyltriiron, tri-Irondodecacarbonyl
Linear Formula:
Fe3(CO)12
CAS Number:
Molecular Weight:
503.66
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Quality Segment
Reagent Type
Core
Showing 1-1 of 1