
Trihexyphenidyl hydrochloride

Synonym(s):
(±)-α-Cyclohexyl-α-phenyl-1-piperidinepropanol hydrochloride, Trihexyphenidyl hydrochloride
Empirical Formula (Hill Notation):
C20H31NO · HCl
CAS Number:
Molecular Weight:
337.93
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing