
Tribasic calcium phosphate

Synonym(s):
Calcium phosphate tribasic, Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate
Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing