
Tetrachlorvinphos

Synonym(s):
Stirofox
Empirical Formula (Hill Notation):
C10H9Cl4O4P
CAS Number:
Molecular Weight:
365.96
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing