
(R)-(−)-O-Acetylmandelic acid

Synonym(s):
(−)-O-Acetyl-D-mandelic acid, (R)-(−)-α-Acetoxyphenylacetic acid
Linear Formula:
CH3CO2CH(C6H5)CO2H
CAS Number:
Molecular Weight:
194.18
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing