Skip to Content
Merck
Pyrrole analytical standard

Pyrrole

Pyrrole analytical standard
Synonym(s):
Azole, Divinylenimine, Imidole, Pyrrole
Empirical Formula (Hill Notation):
C4H5N
CAS Number:
Molecular Weight:
67.09
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing