
Psilocybin-D4 solution

Synonym(s):
Psilocybin-D4
Empirical Formula (Hill Notation):
C12H13D4N2O4P
CAS Number:
Molecular Weight:
288.27
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing