
Progoitrin potassium salt

Synonym(s):
(R)-2-Hydroxy-3-butenylglucosinolate potassium salt, 1-S-[(3R)-3-Hydroxy-N-(sulfooxy)-4-pentenimidoyl]-1-thio-β-D-glucopyranose monopotassium salt, Glucorapiferin potassium salt
Empirical Formula (Hill Notation):
C11H18KNO10S2
CAS Number:
Molecular Weight:
427.49
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing