
Plastic additive 20

Synonym(s):
Oleamide, cis-9,10-Octadecenoamide, cis-9-Octadecenamide, Oleic acid amide
Empirical Formula (Hill Notation):
C18H35NO
CAS Number:
Molecular Weight:
281.48
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing