
Perfluoro-2-propoxypropanoic acid

Synonym(s):
2,3,3,3-Tetrafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propanoic acid, Tetrafluoro-2-(heptafluoropropoxy)propanoic acid
Empirical Formula (Hill Notation):
C6HF11O3
CAS Number:
Molecular Weight:
330.05
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing