
Pentaethylene glycol monodecyl ether

Synonym(s):
C10E5, Decyl pentaethylene glycol ether, Decylpentaglycol, Polyoxyethylene (5) decyl ether
Linear Formula:
CH3(CH2)9(OCH2CH2)5OH
CAS Number:
Molecular Weight:
378.54
Products
Product Category
Formula Weight
Physical Form
Product Line
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing