
N-Decanoyl-N-methylglucamine

Synonym(s):
N-(D-Glucityl)-N-methyldecanamide, N-Decanoyl-N-methyl-D-glucamine, MEGA-10
Empirical Formula (Hill Notation):
C17H35NO6
CAS Number:
Molecular Weight:
349.46
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing