
Methotrexate

Synonym(s):
Methotrexate hydrate, 4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate
Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Physical Form
Activity Spectrum
Agency
Research Area
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
Optical Isomer
Technique
Showing 1-1 of 1