
Methenamine hippurate

Synonym(s):
Hexamethylenetetramine monohippurate
Empirical Formula (Hill Notation):
C6H12N4 · C9H9NO3
CAS Number:
Molecular Weight:
319.36
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing