Skip to Content
Merck
Methacetin-(methoxy-13C) Extra pure, 99 atom % 13C, 99% (CP)

Methacetin-(methoxy-13C)

Methacetin-(methoxy-13C) Extra pure, 99 atom % 13C, 99% (CP)
Synonym(s):
N-(4-Methoxy-13C-phenyl)acetamide
Linear Formula:
CH3CONHC6H4O13CH3
CAS Number:
Molecular Weight:
166.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Special Grade