
Meclozine impurity B

Synonym(s):
4-Chlorobenzhydrol
Linear Formula:
ClC6H4CH(C6H5)OH
CAS Number:
Molecular Weight:
218.68
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing