
Lithium aluminum hydride solution

Synonym(s):
LAH, Lithium alanate, Lithium tetrahydroaluminate
Linear Formula:
LiAlH4
CAS Number:
Molecular Weight:
37.95
Products
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reagent Type
Showing 1-5 of 5
Compare
Product Number
Description
Pricing