
Levothyroxine Sodium Hydrate

Synonym(s):
Levothyroxine sodium hydrate, 3,3′,5,5′-Tetraiodo-L-thyronine monosodium salt hydrate, L-Thyroxin monosodium salt hydrate, Sodium O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine hydrate
Empirical Formula (Hill Notation):
C15H10I4NNaO4 · xH2O
CAS Number:
Molecular Weight:
798.85 (anhydrous basis)
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing