Skip to Content
Merck
Lauric aldehyde ≥95%, stabilized, FCC, FG

Lauric aldehyde

Lauric aldehyde ≥95%, stabilized, FCC, FG
Synonym(s):
Aldehyde C12, Dodecanal, Lauraldehyde, Laurinaldehyde
Linear Formula:
CH3(CH2)10CHO
CAS Number:
Molecular Weight:
184.32
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing