
Hydroxylamine hydrochloride

Synonym(s):
Hydroxylammonium chloride
Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing