
Heptakis(2,3,6-tri-O-methyl)-β-cyclodextrin

Synonym(s):
2,3,6-Tri-O-methyl-β-cyclodextrin, Trimethyl-β-cyclodextrin
Empirical Formula (Hill Notation):
C63H112O35
CAS Number:
Molecular Weight:
1429.54
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing