
Esculetin

Synonym(s):
6,7-Dihydroxycoumarin, Cichorigenin, Esculetin
Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing