
Disopyramide

Synonym(s):
α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide
Empirical Formula (Hill Notation):
C21H29N3O
CAS Number:
Molecular Weight:
339.47
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing