
Digitonin

Synonym(s):
Digitin
Empirical Formula (Hill Notation):
C56H92O29
CAS Number:
Molecular Weight:
1229.31
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Tissue
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing