
Coptisine Chloride

Synonym(s):
Coptisine chloride, 6,7-Dihydrobis[1,3]benzodioxolo[5,6-a:4‘,5‘-g]quinolizinium chloride, Coptisine hydrochloride
Empirical Formula (Hill Notation):
C19H14ClNO4
CAS Number:
Molecular Weight:
355.77
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing