
cis-Bis(2,2′-bipyridine)dichlororuthenium(II) hydrate

Empirical Formula (Hill Notation):
C20H16Cl2N4Ru · xH2O
CAS Number:
Molecular Weight:
484.34 (anhydrous basis)
Products
Brand
Formula Weight
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing