
cis-1,2,3,6-Tetrahydrophthalimide

Synonym(s):
cis-4-Cyclohexene-1,2-dicarboximide
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2