
CHAPSO

Synonym(s):
3-([3-Cholamidopropyl]dimethylammonio)-2-hydroxy-1-propanesulfonate, CHAPSO
Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-4 of 4