
Candesartan cilexetil Related Compound B

Synonym(s):
2,3-Dihydro-2-oxo-3-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-benzimidazole-4-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester
Empirical Formula (Hill Notation):
C31H30N6O6
CAS Number:
Molecular Weight:
582.61
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2