
Betadex

Synonym(s):
β-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin
Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing