
Azocarmine G

Synonym(s):
Acid Red 101, Rosinduline
Empirical Formula (Hill Notation):
C28H18N3NaO6S2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing