
Anti-OTC antibody produced in rabbit

Synonym(s):
Anti-OTCase, Anti-Ornithine carbamoyltransferase, mitochondrial precursor, Anti-Ornithine transcarbamylase, Anti-OCTD, Anti-Ornithine carbamoyltransferase
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing