Skip to Content
Merck
Amiodarone Related Compound H United States Pharmacopeia (USP) Reference Standard

Amiodarone Related Compound H

Amiodarone Related Compound H United States Pharmacopeia (USP) Reference Standard
Synonym(s):
2-Chloro-N,N-diethylethylamine hydrochloride, 2-(Diethylamino)ethyl chloride hydrochloride, 2-Chlorotriethylamine hydrochloride
Linear Formula:
(C2H5)2NCH2CH2Cl · HCl
CAS Number:
Molecular Weight:
172.10

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing