
(Aminomethyl)polystyrene

Synonym(s):
AM-polystyrene, Benzylamine polymer-bound, Poly(styrene-co-divinylbenzene), aminomethylated, Poly(styrene-co-vinylbenzylamine-co-divinylbenzene, Polystyrene crosslinked with divinylbenzene, aminomethylated
Products
Brand
Physical Form
Quality Segment
Functional Group
Application
Particle Size (µm)
Chain Type
Reaction Type
Reactivity
Showing 1-6 of 6