
6-Carboxy-tetramethylrhodamine N-succinimidyl ester

Synonym(s):
"5-Carboxytetramethylrhodamine N-succinimidyl ester"
Empirical Formula (Hill Notation):
C29H25N3O7
CAS Number:
Molecular Weight:
527.52
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing