
3-Hydroxytyrosol

Synonym(s):
2-(3,4-Dihydroxyphenyl)ethanol, 3,4-Dihydroxyphenethyl alcohol, DOPET, Homoprotocatechuyl alcohol
Empirical Formula (Hill Notation):
C8H10O3
CAS Number:
Molecular Weight:
154.16
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing