![3-Hydroxyglutaric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/194/058/13a6b49c-b75b-4be2-a42b-781f88a2d6d7/640/13a6b49c-b75b-4be2-a42b-781f88a2d6d7.png)
3-Hydroxyglutaric acid
![3-Hydroxyglutaric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/194/058/13a6b49c-b75b-4be2-a42b-781f88a2d6d7/640/13a6b49c-b75b-4be2-a42b-781f88a2d6d7.png)
Synonym(s):
β-Hydroxyglutaric acid, 3-Hydroxypentanedioic acid
Empirical Formula (Hill Notation):
C5H8O5
CAS Number:
Molecular Weight:
148.11
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2