![3-Hydroxybutyric acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/103/209/1894a759-daa8-4931-9169-8aacf41733fa/640/1894a759-daa8-4931-9169-8aacf41733fa.png)
3-Hydroxybutyric acid
![3-Hydroxybutyric acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/103/209/1894a759-daa8-4931-9169-8aacf41733fa/640/1894a759-daa8-4931-9169-8aacf41733fa.png)
Synonym(s):
(±)-3-Hydroxybutanoic acid, DL-β-Hydroxybutyric acid
Linear Formula:
CH3CH(OH)CH2COOH
Molecular Weight:
104.10
EC Number:
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing