Product Name
Poly(ethylene glycol) average Mn 600, waxy solid (moist)
Product Number
202401
Product Brand
ALDRICH
CAS Number
25322-68-3
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless and White to Off White |
APPEARANCE (FORM) | Viscous Liquid or Semi-Solid or Paste or Solid |
PH-TEST | 4.5 - 7.5 |
PH (METHOD) | C = 5 %, H2O |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |
Remark | MOLECULAR NUMBER: 570 - 630 |