Skip to Content
Merck
All Photos(1)

Key Documents

BP858

Dibenzosuberone

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-Dibenzosuberone, 10,11-Dihydro-5H-dibenzo[a,d]cyclohepten-5-one, Dibenzocycloheptadienone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O
CAS Number:
Molecular Weight:
208.26
Beilstein:
1454355
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

nortriptyline, amitriptyline

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

refractive index

n20/D 1.6332 (lit.)

bp

148 °C/0.3 mmHg (lit.)

mp

32-34 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O=C1c2ccccc2CCc3ccccc13

InChI

1S/C15H12O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8H,9-10H2

InChI key

BMVWCPGVLSILMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopoeia.

Application

Dibenzosuberone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Amitriptyline Oral Solution
  • Amitriptyline Tablets
  • related product

    Product No.
    Description
    Pricing

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    235.4 °F - closed cup

    Flash Point(C)

    113 °C - closed cup


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service