Skip to Content
Merck
All Photos(2)

Key Documents

29538-U

Supelco

Empty Tube for Thermal Desorption

greener alternative

glass, O.D. x I.D. x L 1/4 in. × 3 1/2 in., fritted, pkg of 10 ea

Synonym(s):

empty glass Thermal Desorption Tube (fritted)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Empty Tube for Thermal Desorption, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., pkg of 10 ea

material

glass TD tube

Quality Level

feature

fritted

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Empty tube for thermal desorption is a glass thermal desorption tube available for packing adsorbents, or sample preparation applications.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application


  • Impact of thermal desorption tubes on the variability of exhaled breath data.: This study investigates the impact of using thermal desorption tubes on the variability of data collected from exhaled breath analysis, highlighting the importance of consistent sampling methods for accurate analytical results (Jung et al., 2023).

  • Concentrically packed high flow air sampler for parts-per-trillion volatile and semi-volatile organic compounds.: The research introduces a high-flow air sampler designed to detect volatile and semi-volatile organic compounds at extremely low concentrations, demonstrating the efficiency of thermal desorption in air quality monitoring (Xie et al., 2017).

  • Performance test of a sorbent tube sampler with respect to analyte loss in collecting biogenic volatile organic compounds.: This paper evaluates the performance of sorbent tube samplers, focusing on analyte loss and its implications for the accurate measurement of biogenic volatile organic compounds (Ullah and Kim, 2015).

  • Extent of sample loss on the sampling device and the resulting experimental biases when collecting volatile fatty acids (VFAs) in air using sorbent tubes.: The study examines sample loss and experimental biases associated with using sorbent tubes for collecting VFAs in air, emphasizing the need for optimized sampling protocols (Kim and Kim, 2013).


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service