Skip to Content
Merck
All Photos(1)

Key Documents

SML3990

Sigma-Aldrich

BODIPY FL-NHS ester

new

≥98% (HPLC)

Synonym(s):

(T-4)-[1-[3-[5-[(3,5-Dimethyl-2H-pyrrol-2-ylidene-κN)methyl]-1H-pyrrol-2-yl-κN]-1-oxopropoxy]-2,5-pyrrolidinedionato]difluoroboron, BDP FL NHS ester, BDP FL SE, BODIPY FL NHS, Bodipy FL succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18BF2N3O4
CAS Number:
Molecular Weight:
389.16
UNSPSC Code:
12352107
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Light orange to very dark red

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CCC1=CC=C2C=C3C(C)=CC(C)=[N]3[B+3]([F-])([N-]21)[F-])ON4C(CCC4=O)=O

Biochem/physiol Actions

Amine reactive green fluorescent dye that can be used to label molecules like antibodies, proteins, peptides, and nucleic acids.



BODIPY FL-NHS ester is a versatile amine sensitive fluorescent labeling reagent that can be used to tag antibodies, proteins, peptides, and nucleic acids. BODIPY (boron-dipyrromethene) dyes are known for their excellent photophysical properties, including high fluorescence quantum yield, good photostability, and wide spectral tunability. BODIPY FL has a distinctive absorption peak (excitation) at 503-505 nm and an emission peak at 512-514 nm, resulting in bright green fluorescence. BODIPY FL is used in a wide range of applications, including immunofluorescence, live cell imaging, FRET experiments, and other fluorescence-based studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengrong Pei et al.
Bioconjugate chemistry, 34(12), 2194-2204 (2023-09-25)
α6β4* nicotinic acetylcholine receptor (nAChR) (* represents the possible presence of additional subunits) is mainly distributed in the central and peripheral nervous system and is associated with neurological diseases, such as neuropathic pain; however, the ability to explore its function
Jenna M Franke et al.
Journal of the American Chemical Society, 141(32), 12824-12831 (2019-07-25)
Fluorophores based on the BODIPY scaffold are prized for their tunable excitation and emission profiles, mild syntheses, and biological compatibility. Improving the water-solubility of BODIPY dyes remains an outstanding challenge. The development of water-soluble BODIPY dyes usually involves direct modification
Jun Yang et al.
The Journal of pharmacology and experimental therapeutics, 321(2), 462-468 (2007-02-10)
Despite the widely accepted assumption that most endosomal compartments are acidic, evaluation of the efficiency of pH-dependent drug release from a ligand-targeted drug conjugate during receptor-mediated endocytosis is lacking. Therefore, we have characterized the kinetics of pH-dependent drug release from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service