Skip to Content
Merck
All Photos(1)

Key Documents

13118

Sigma-Aldrich

Magnesium carbonate basic

purum, light, ≥40% Mg (as MgO) basis, powder (light)

Synonym(s):

Magnesium hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCO3
CAS Number:
Molecular Weight:
84.31
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
NA.22

Quality Level

grade

purum

form

powder (light)

composition

Mg (as MgO), ≥40%

impurities

≤0.005% heavy metals (as Pb)

anion traces

chloride (Cl-): ≤500 mg/kg

cation traces

Ca: ≤1%
Fe: ≤500 mg/kg

SMILES string

[Mg++].O[Mg]O.[O-]C([O-])=O

InChI

1S/CH2O3.2Mg.2H2O/c2-1(3)4;;;;/h(H2,2,3,4);;;2*1H2/q;2*+2;;/p-4

InChI key

UDYRPJABTMRVCX-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used as:
  • A stabilizer for phase formation
  • IR high-pass transmittance filter

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M. G. Baldecchi and B. Melchiorri,
Infrared Physics, 13, 189-211 (1973)
J. Park, et al
J. Mater. Sci., 33, 5671-5675 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service