Skip to Content
Merck
All Photos(1)

Key Documents

1.04763

Supelco

Iodine

99.999 Suprapur®

Synonym(s):

Iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
MDL number:
UNSPSC Code:
12141916
EC Index Number:
231-442-4
NACRES:
NA.21

vapor pressure

0.41 hPa ( 25 °C)

Quality Level

Assay

≥99.8% (iodometric)

form

solid

potency

14000 mg/kg LD50, oral (Rat)
1425 mg/kg LD50, skin (Rabbit)

pH

5.4 ( in H2O, saturated solution)

kinematic viscosity

0.57 cSt(116 °C) (liquid)

bp

184.4 °C/363.9 °F (at 1,013 hPa)

mp

113.5 °C (236.3 °F)

solubility

0.3 g/L °C ((77 °F) - slightly soluble in water)

density

4.93 g/cm3 at 20 °C (68 °F)

bulk density

2100 kg/m3

storage temp.

15-25°C

SMILES string

II

InChI

InChI=1S/I2/c1-2
1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N
PNDPGZBMCMUPRI-UHFFFAOYSA-N

Application



  • Iodine-Promoted Reductive Sulfenylation Using Ketones as Hydride Donors.: The research demonstrates the utility of iodine in promoting reductive sulfenylation reactions with ketones, offering a novel method for synthesizing sulfenylated compounds (Duan et al., 2024).


  • I(2)/DMSO-mediated oxidative C-C and C-heteroatom bond formation: a sustainable approach to chemical synthesis.: The research introduces a sustainable method for C-C and C-heteroatom bond formation using iodine and DMSO, enhancing green chemistry applications (Singhal et al., 2024).



Analysis Note

Purity (metallic): ≥ 99.999 %
Assay (iodometric): ≥ 99.8 %
Chlorine and Bromine (as Cl): ≤ 10 ppm
Ag (Silver): ≤ 0.010 ppm
Al (Aluminium): ≤ 0.050 ppm
Au (Gold): ≤ 0.050 ppm
Ba (Barium): ≤ 0.050 ppm
Be (Beryllium): ≤ 0.005 ppm
Bi (Bismuth): ≤ 0.050 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.010 ppm
Fe (Iron): ≤ 0.050 ppm
Ga (Gallium): ≤ 0.050 ppm
In (Indium): ≤ 0.010 ppm
K (Potassium): ≤ 0.50 ppm
Li (Lithium): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.050 ppm
Mn (Manganese): ≤ 0.005 ppm
Mo (Molybdenum): ≤ 0.020 ppm
Na (Sodium): ≤ 0.10 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.050 ppm
Sn (Tin): ≤ 0.050 ppm
Sr (Strontium): ≤ 0.010 ppm
Ti (Titanium): ≤ 0.050 ppm
Tl (Thallium): ≤ 0.050 ppm
V (Vanadium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service