Key Documents
SYNP170001
Synthware round bottom pressure vessel with PTFE bushing
75 mL, O.D. × L 46 mm × 129 mm, #15 bushing
Synonym(s):
PTFE Bushing Vessel, Pressure Vessel with Bushing, Round Bottom Pressure Vessel, Synthware P170001, pressure tube
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware P170001
O.D. × L
46 mm × 129 mm
capacity
75 mL
75 mL
fitting
#15 bushing
Related Categories
General description
Heavy wall round bottom vessels have an internal thread for use with a 15 mm PTFE bushing as a pressure seal. This unique design allows hand tightening and loosening for easy access into the vessel. The O-ring is positioned on the bushing above the thread for improved sealing. The vessels are useful for pressure reactions and applications such as polymerization and analysis of halogens by thermal decomposition in the presence of lime. Complete apparatus is supplied with vessel, PTFE bushing and Viton® O-ring. *Important Warnings for Pressurized Glassware: Due to varying conditions, we cannot guarantee glass vessels from breakage under pressure. Always work behind a safety shield.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company