Comparison of Granulated and Powdered Dehydrated Culture Media
Dehydrated culture media (DCM) for the growth of microorganisms in broth or on agar plates are available in the market in both powdered and granulated forms. Though their chemical composition is identical, they differ with respect to their particle sizes. Traditional powdered media consist of tiny particles with a size less than 100 µm, but the granules are visible to the naked eye, reaching 1,000 µm or more. Both types of dehydrated culture media offer identical growth promotion performance. Let’s compare and find out other differences between these two types of dehydrated culture media.
Figure 1.Granulated (left) and powdered (right) microbial culture media.
Watch the video to learn about the many advantages of using granulated dehydrated culture media for easy and mess-free media preparation.
FROM POWDERED MEDIA TO GRANULES
Dehydrated culture media granules are firm but loosely bound powdered media. To produce them, the powder mixture is subjected to a water- and additive-free roller compaction process known as granulation. This process, which is purely mechanical and also suitable to heat-sensitive components, leads the powdered media to clump and form granules of different sizes (Figure 2) that dissolve rapidly in water. During product transportation, a small proportion of the granules fragments into smaller ones and abrasion can lead to powder settling at or near the bottom of the container. Studies have shown that these powder residuals are identical to the granules and offer the same growth-promoting performance.1
Figure 2.Example of the particle frequency in a container of agar medium.
Selective ingredients can be incorporated into granules just as well as into powdered media, freeing labs of the need to purchase and store expensive supplements.
ADVANTAGES OF GRANULATED CULTURE MEDIA
In the 1950s we pioneered the granulation of powdered dehydrated culture media. Since then, our granulated media, offered under the GranuCult® brand with the addition “plus” or “prime” to indicate the QC level, offers many advantages over powdered media. Although granulation is an additional step in the manufacturing process, the extra costs have not posed an obstacle because the media are produced on an industrial scale, keeping the overheads low. Most of our dehydrated culture media to prepare agar plates or broths for microbiological quality control testing of pharmaceuticals, food, and beverages are now available in the granulated form.
Granulated media offer advantages over powdered media with regard to:
Safety: Better protection for lab staff
When powdered media are weighed and transferred into flasks, fine particles drift as visible clouds through the air and across the working area, causing laboratory personnel to inhale them. Because many culture media contain hazardous or toxic substances (Table 1), this causes many health concerns. Bile salts, for example, constituents of Violet Red Bile Glucose Agar, irritate the mucosa and trigger coughing. Powdered media particles settling on the skin, eyes, and ears can lead to allergic reactions.
Granulated dehydrated culture media, on the other hand, stir up much less powder during handling. Consequently, the danger of inhaling hazardous or toxic substances is significantly lower, and this helps to maintain a safer, cleaner working environment.
Accuracy: No separation or weighing issues
Granulation preserves the homogeneity of culture media by capturing its original component distribution in each granule and by preventing those components from settling into separate layers. This eliminates the need for tumbling to redistribute components, as is sometimes required with powdered media. In addition, handling and weighing powdered media can lead to dust settling on and around balances, causing not only the need for extra cleaning but also the risk of inaccurate weighing results.
Figure 3.Dust formation of granulated (left) and powdered (right) culture medium after filling in an Erlenmeyer flask.
Speed: Granules dissolve faster
Weighing and preparing granulated media is easy thanks to its free-flowing properties, and this takes less time than with powdered media. After adding water the granules dissolve easily within minutes (except agar and gelatin, of course). This is because water coats and penetrates the granules much quicker than it does fine powders. Powders tend to float on the water until forced into solution, while granules sink into the water naturally, beginning to dissolve as soon as they are coated with water.
Figure 4.Dissolving of granulated (left) and powdered (right) culture media in water.
Convenience: No clumping, no sticking
Powdered dehydrated culture media tends to clump and stick to the bottom of culture media flasks. Such clumps are sometimes difficult to dissolve. Powdered media also likes to stick to a flask’s neck, making cleaning necessary as undissolved media can caramelize upon subsequent heating.
Granulated media, on the other hand, possesses better flow properties, making it easy to weigh out and transfer from the weighing booth to a flask. It doesn’t stick to the neck of the flask or clump within it. Swirling the flask gently is enough to dissolve the media.
Figure 5.Solubility of granulated (left) and powdered (right) culture media after stirring.
Stability: Longer shelf life
Most of our granulated dehydrated culture media have a shelf life of 5 years, much longer than some commercial powdered media with a 3-year shelf life. Granulated media have a lower water content when they are produced, and the granules themselves are less affected by ambient moisture when containers are opened and closed. There are less chances of clumping even under warm or humid conditions, ensuring a stable product during its shelf life. Longer shelf lives generally mean that less media has to be thrown away as a consequence of product expiry, which saves costs over time.
Conclusion: Granulated Culture Media Vs Powdered Media
While granulated dehydrated culture media possess the same growth promotion properties as powdered media, they are at a clear advantage in terms of safety, accuracy, speed, convenience, and stability. Because the purely mechanical process of granulating powdered media involves neither adding substances nor causing damage to heat-sensitive ingredients, it has been possible to apply it to most culture media used in microbiological quality control testing of food, beverages, and pharmaceuticals.
Explore our extensive portfolio of microbial culture media raw materials , which includes both granulated and powdered biological raw materials. Our offerings feature dehydrated raw materials such as peptones, agar-agar, and extracts, along with various additives. All our raw materials are designed for safe handling and optimal performance, backed by stringent quality control and assurance standards throughout the production process. Visit our webpage.