HPLC Analysis of Six Enkephalin Peptides (Chiral and Achiral) on Astec® CHIROBIOTIC® T2
Materials
analytical column
CONDITIONS
column
CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 μm particles (16024AST)
mobile phase
[A] 5 mM ammonium formate, pH 3.3; [B] acetonitrile; (25:75, A:B)
flow rate
0.5 mL/min
detector
UV, 230 nm
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC