HPLC Analysis of Miconazole Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP
CONDITIONS
column
CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase
[A] acetonitrile; [B] 20 mM ammonium acetate, pH 4.0; (25:75, A:B)
flow rate
0.6 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
5 μL
sample
miconazole, 1 mg/mL in acetonitrile:water (50:50)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null